Clamp, Three-Prong, Vertical, Ace Glass
Supplier: Ace Glass
11077-18
89083-210EA 88.74 USD
89083-210
Clamp, Three-Prong, Vertical, Ace Glass
Clamps General Purpose Clamps
This three-prong grip clamp is designed to hold dispensing burets, thermometers, reflux condensers, and other laboratory glassware, in a vertical position
The vinlylized jaws will hold objects up to 57mm in diameter.