Adapter, Straight Connecting with Easy-Action Stopcock, Ace Glass Incorporated
Supplier: Ace Glass
3844-05 3844-31 3844-07 3844-34 3844-47 3844-09 3844-36 3844-49
89082-722EA 282.37 USD
89082-722 89082-724 89082-754 89082-756 89082-758 89082-784 89082-786 89082-788
Adapter, Straight Connecting with Easy-Action Stopcock, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This straight connecting adapter has an Easy-Action threaded plug.
  • Easy-Action threaded plug
  • [ST] inner and reinforced outer joints
  • Fabricated from borosilicate glass
Adapter features standard taper inner and reinforced outer joints at top and bottom.