Adapter, Straight Connecting PTFE Stopcock, Ace Glass Incorporated
Supplier: Ace Glass
3847-12 3847-08 3847-10 3847-04 3847-06
89082-728EA 295.24 USD
89082-728 89082-744 89082-774 89082-794 89082-796
Adapter, Straight Connecting PTFE Stopcock, Ace Glass Incorporated
Adapters Adapters, Conical Ground Joints
This adapter comes with a PTFE plug stopcock and o-ring top joint, and either a standard taper or spherical joint at the bottom.
  • PTFE plug stopcock
  • O-ring top joint
  • Either [ST] or spherical bottom joint
  • Fabricated from borosilicate glass
Adapter features a 4mm PTFE plug stopcock and #15 o-ring top joint.
O-ring is made of FETFE. FETFE is an exclusive compound which is a fluoroelastomer with TFE additives. It has good compression set along with excellent temperature and chemical compatibility. USP Class VI material.
Ordering information: Comes complete with 89061-002 FETFE o-ring for top joint.