Adapter, Straight Connecting with Glass Stopcock, Ace Glass Incorporated
Supplier: Ace Glass
3846-13 3846-09 3846-11 3846-05 3846-07
89082-726EA 308.05 USD
89082-726 89082-742 89082-772 89082-790 89082-792
Adapter, Straight Connecting with Glass Stopcock, Ace Glass Incorporated
Adapters Adapters, Conical Ground Joints
This adapter comes with a glass plug stopcock and o-ring top joint, and either a standard taper or spherical joint at the bottom.
  • Glass plug stopcock
  • O-ring top joint
  • Either [ST] or spherical bottom joint
  • Fabricated from borosilicate glass
Adapter features a 4mm glass plug stopcock and #15 o-ring top joint.
Top o-ring is made of FETFE. FETFE is an exclusive compound which is a fluoroelastomer with TFE additives. It has good compression set along with excellent temperature and chemical compatibility. USP Class VI material.
Ordering information: Comes complete with 89061-002 FETFE o-ring for top joint.