Adapter, Bellows, PTFE, Ace Glass Incorporated
Supplier: Ace Glass
13441-23 13441-28 13441-32 13441-36 13441-19
89082-658EA 226.16 USD
89082-658 89082-660 89082-662 89082-664 89082-666
Adapter, Bellows, PTFE, Ace Glass Incorporated
Adapters Adapters, Conical Ground Joints
This bellows-type adapter is constructed from PTFE.
  • Safe for use in temperatures up to 200°C
  • Made of polytetrafluoroethylene
Adapter is used to correct the alignment of standard taper joints and relieve stress in reaction systems.