Adapter, Offset, Ace Glass Incorporated
Supplier: Ace Glass
5268-10 5268-15 5268-21 5268-35 9089-08 9089-40 5268-54 5268-56 5268-58 5268-114 5268-124 5273-02 5273-04 5273-114 5273-124
89051-532EA 82.75 USD
89051-532 89051-534 89051-536 89051-538 89064-868 89082-644 89082-646 89082-648 89082-650 10098-744 10098-746 10098-750 10098-752 10098-754 10098-756
Adapter, Offset, Ace Glass Incorporated
Adapters Adapters, Conical Ground Joints
Offest adapters are used to gain better vertical clearance when using addition funnels, condensers, or other apparatus.
  • Bottom inner and top outer joints are slightly offset
  • Made of borosilicate glass
Both bottom inner and top outer joints are slightly offset from each other. The 10° angled offset adapters are designed for use with reactor heads that have 10° angled side necks for transitioning to 90° vertical.