Adapter, Hose Connection, Ace Glass Incorporated
Supplier: Ace Glass
5218-10
89082-642EA 46.02 USD
89082-642
Adapter, Hose Connection, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This adapter features an o-ring joint at one end and a hose connection at the other end.
  • O-ring joint
  • With hose connection
  • Fabricated from borosilicate glass
Adapter has a #15 o-ring joint at one end and a hose connection at the other end. Use with a 3/8-inch or 7/16-inch I.D. tubing, size E hose connection.