Hose Connection Adapter, [ST], with Frit, Ace Glass Incorporated
Supplier: Ace Glass
5216-110 5216-116 5216-118 9069-115 9069-116
89082-624EA 71.48 USD
89082-624 89082-626 89082-628 89082-636 89082-638
Hose Connection Adapter, [ST], with Frit, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
These adapters feature a straight hose connection at top and contain a fritted disc.
  • Bottom ground [ST] inner joint
  • Top straight hose connection
  • Fritted disc
  • Fabricated from borosilicate glass
Featuring a ground standard taper inner joint at the bottom and a straight hose connection at the top. Adapters contain an integral Porosity A (145–174 micron) fritted disc.