Adapter, Connecting, Ace Glass Incorporated
Supplier: Ace Glass
5129-11 5129-15 5129-25 5129-07
89082-504EA 124.12 USD
89082-504 89082-506 89082-508 89082-510
Adapter, Connecting, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This adapter is used with vacuum manifolds as port connections.
  • O-ring joint on one end
  • Ace-Thred on opposite end
  • One o-ring included
  • Fabricated from borosilicate glass
Featuring a #15 o-ring joint on one end with an Ace-Thred on the opposite end. Supplied with one o-ring.