Adapter, Adjustable Electrode, with Joint, Ace-Thred, Ace Glass Incorporated
Supplier: Ace Glass
5038-04 5038-05 5038-06 5038-07 5038-16 5038-17 5038-26 5038-27 5038-08 5038-28
89051-226EA 90.24 USD
89051-226 89051-228 89051-230 89051-232 89051-234 89051-236 89051-238 89051-240 89082-482 89082-558
Adapter, Adjustable Electrode, with Joint, Ace-Thred, Ace Glass Incorporated
Adapters Adapters, Conical Ground Joints
This Ace-Thred adapter is used with platinum or other wire electrodes that normally would be step sealed to glass.
  • Electrode may be adjusted or removed
  • Threaded bushing
  • Silicone rubber septum
  • Bottom ground [ST] inner joint
  • Fabricated from borosilicate glass
With threaded bushing and silicone rubber septum for sealing, the electrode may be adjusted for depth or removed if desired. Has ground standard taper inner joint at bottom. Glass capillary is 1-1/2 to 2mm with 76mm of stem for sealing to apparatus. Size is determined by bore of hole in bushing. Over length of glass is 140mm.
Ordering information: Available complete with 89077-538 silicone rubber septum and nylon bushing, or as glassware only.