Receiving Tube/Cylinder, Ace Glass Incorporated
Supplier: Ace Glass
8387-04
89082-460EA 265.34 USD
89082-460
Receiving Tube/Cylinder, Ace Glass Incorporated
Tubes Test Tubes
This graduated, tapered bottom, receiving tube is a replacement for receivers on all glassware sets supplied with Heidolph® LR4000 Series rotary evaporators or with other evaporators.
  • The 100 ml size is graduated in 0.5 ml increments from 0 to 10 ml, and in 5 ml increments from 10 to 100 ml.
Joint is spherical 35/20 socket. Fabricated from borosilicate glass.