ART® 20P Self-Sealing Barrier Pipette Tips, Molecular BioProducts
Supplier: Thermo Scientific
2149PIW 2149P 2149P-05 2149P-HR 2149P-05-HR 2149P-RI 2149P-05-RI
53225-764CS 284.82 USD
53225-764 72830-046 72830-440 89082-074 89082-076 89082-110 89082-112
ART® 20P Self-Sealing Barrier Pipette Tips, Molecular BioProducts
Pipette Tips
Designed with a patented, self-sealing barrier, ART tips are the only tips that completely block the passage of aqueous based liquids and aerosols from the sample to the pipettor.
  • All tips are pre-sterilized
Low retention tips reduce sample loss due to adhesion, allowing for more accurate dispensing and an increased level of reproducibility.
The purity of ART tips is achieved by constant control against contamination from any source. Electron beam sterilization is used because it ensures total disruption of any molecules amplifiable by PCR. ARTMore Product Information