Recycle-Pak® Lamp Recycling Kit, Bulbtronics
Supplier: Avantor
89079-086CS 24755.21 USD
89079-086 89079-092
Recycle-Pak® Lamp Recycling Kit, Bulbtronics
Lights Fluorescent Lights, Lamps and Bulbs
144 Lamp recycling kit is applicable for straight linear fluorescent and High Intensity Discharge (HID) lamps. This takes up to 3700 HID lamps or 1800 4' T12 and T8 fluorescent tubes. It is mainly for large quantity recycling.
044 Lamp recycling kit (8') takes 15 T12 or 30 T8 lamps. Do not place broken lamps into container; intact bulbs only.