di-Sodium tetraborate decahydrate ≥99% NF suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

Synonyms: Borax tetra, Sodium tetraborate decahydrate, Sodium Borate decahydrate

S1183-50KGBL S1183-12KG S1183-500GM S1183-2.5KG
89079-002EA 2449.8 USD
89079-002 700000-414 95035-736 95035-734
di-Sodium tetraborate decahydrate ≥99% NF suitable for use as excipient
di-Sodium tetraborate decahydrate

Sodium Borate, Decahydrate, NF is a pH control buffering excipient. 


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: Na₂B₄O₇·10H₂O
Boiling Pt: 320 °C (1013 hPa)
Melting Pt: 75 °C
Density: 1.73 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149193
CAS Number: 1303-96-4
EINECS: 215-540-4
REACH: 01-2119490790-32
Merck Index: 13,08662

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR