Polyvinylpyrrolidone, M.W. ∼ 40,000 K-30 USP suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Povidone, Polyvinylpyrrolidon, Polyvidone, PVP

P1454-5KG P1454-100GM P1454-500GM P1454-5GM P1454-12KGBL P1454-50KGBL
700008-862EA 2488.1 USD
700008-862 95035-244 95035-246 95035-248 89078-978 89078-980
Polyvinylpyrrolidone, M.W. ∼ 40,000 K-30 USP suitable for use as excipient
Polyvinylpyrrolidone

Melamine statement (available upon request) BSE/TSE statement (available upon request)


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: (C₆H₉NO)n
Storage Temperature: Ambient
MDL Number: MFCD01076626
CAS Number: 9003-39-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR