Peppermint oil NF
Supplier: Spectrum Chemical Mfg. Corp.
|
|
Peppermint Oil, NF - Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities
GHS hazard : Exclamation mark
Storage Temperature:
Ambient |
MDL Number:
MFCD00147870 CAS Number: 8006-90-4 |
Specification Test Results
Esters(As Menthyl Acetate) | 5.0 % min. |
Total Menthol(Free and As Esters) | 50.0 % min. |
Specific Gravity @ 25°C | 0.896 - 0.908 |
Angular Rotation | -18° to -32° |
Refractive Index @ 20°C | 1.459 - 1.465 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...