Isopropyl palmitate ≥90% NF

Supplier: Spectrum Chemical Mfg. Corp.

IS160-4LTGL IS160-500CS6 IS160-20LTMT IS160-4CS4 IS160-500MLG IS160-500MLG IS160-200LTBL
95033-744EA 479.08 USD
95033-744 95033-746 700005-010 95033-742 700005-012 95033-748 89078-816
Isopropyl palmitate ≥90% NF
Isopropyl palmitate

Isopropyl Palmitate, NF is derived from palm oil and is used as a thickening agent and antistatic agent. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: C₁₉H₃₈O₂
Boiling Pt: 160 °C (2 mmHg)
Melting Pt: 13.5 °C
Density: 0,855 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008993
CAS Number: 142-91-6
EINECS: 205-571-1

Order Now

Specification Test Results

Assay 90% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR