Peptide Vessel Shaker, 240 V
Supplier: Ace Glass
13814-44
89078-084EA 4569.47 USD
89078-084
Peptide Vessel Shaker, 240 V
Shakers and Mixers Orbital Shakers
This is a 180 degree wrist type action shaker that simulates shaking by hand, agitation rate can be varied from 2 to 30 rpm.
  • 240 V model
Will shake two flasks simultaneously. Two laboratory clamps hold glassware securely. Shaft bearing permits operation in a cold room or incubator. DC filtering allows the motor to run cooler and promotes longer motor life. Base configuration prevents walking. Fuse protected, with on/off switch, variable speed control knob and six foot 3-wire grounded cord.
Caution: 240 V model. Corresponding model with voltage suitable for use in US (89078-082) is also available, please see separate product page.