Peptide Vessel Shaker, Ace Glass Incorporated
Supplier: Ace Glass
13814-20
89078-082EA 4278.03 USD
89078-082
Peptide Vessel Shaker, Ace Glass Incorporated
Shakers and Mixers Orbital Shakers
This is a 180 degree wrist type action shaker that simulates shaking by hand, agitation rate can be varied from 2 to 30 rpm.
Will shake two flasks simultaneously. Two laboratory clamps hold glassware securely. Shaft bearing permits operation in a cold room or incubator. DC filtering allows the motor to run cooler and promotes longer motor life. Base configuration prevents walking. Fuse protected, with on/ off switch, variable speed control knob and six foot 3-wire grounded cord