Glas-Col Laboratory Stirrers, Ace Glass Incorporated
Supplier: Ace Glass
13647-14 13647-30 13647-35
89077-798EA 2900.51 USD
89077-798 89077-800 89077-802
Glas-Col Laboratory Stirrers, Ace Glass Incorporated
Stirrers Overhead Stirrers
This stirrer is recommended for heavy mixtures of high viscosity
Single shafted, direct drive totally enclosed 1/17 hp. cool, permanent-magnet, direct drive or geared motor. Controller: Continuously variable solid state speed controller. Complete item includes motor, mounting rod, controller, stainless steel paddle.