Dual Motor Speed and Power Controller, Solid State, Ace Glass Incorporated
Supplier: Ace Glass
13532-10
89077-718EA 1393.31 USD
89077-718
Dual Motor Speed and Power Controller, Solid State, Ace Glass Incorporated
Controllers Utility Controllers
Comes with a buffered load control and DC filtration to provide more torque at higher speeds and a higher top speed than motor rating
A compact 8.9 x 8.9 x 17.8 cm (31/2 x 31/2 x 7"), lightweight 1 Kg. (21/4 lbs.) solid state control with rugged control regulation and rectification circuitry. Two output sockets AC and DC which work in conjunction. Fwd-Off-Rev. DC control switch only. Fused for AC and DC outputs, 10 amps and three (3) amps respectively. Rear ring stand clamp for easy mounting and access. Light blue modern type case with protective finish. Red pilot light. Large dial plateMore Product Information