Omnifit Connector Bodies, High Pressure, Ace Glass Incorporated
Supplier: Ace Glass
12741-04 12741-25 12741-26 12741-40
89077-480EA 60.14 USD
89077-480 89077-482 89077-484 89077-486 89077-488 89077-490 89077-492 89077-498
Omnifit Connector Bodies, High Pressure, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
Ominfit connector bodies are made exclusively of virgin PTFE for maximum chemical inertness.
  • No dead spaces within internal passages
  • Tefzel threaded couplings
  • Rated at 1000psi
  • Autoclavable
The internal passages are 0.8 mm I.D. throughout and there are no dead spaces within. The connectors have 1/4"-28 threaded couplings of Tefzel for instant connection to tube end fittings, plugs or adapters. The connectors are rated at 1000psi and may be autoclaved.