Brady® i5100 IP 1 Core R6400 Series Printer Ribbon
Supplier: Brady Worldwide
IP-R6400
89077-381RL 161.31 USD
89077-381
Brady® i5100 IP 1 Core R6400 Series Printer Ribbon
Printer Ribbons
IP-Enabled ribbons come spooled on a specially designed core that includes a Smart Cell RFID tag that communicates with the Brady IP® industrial label printer.
  • Chemical-resistant
  • Abrasion-resistant
One-color ribbon for printing a single color onto a label. R6400 Series Resin Formulation offers superior chemical and abrasion resistance when printed on the appropriate label material from Brady®. Recommended for use with materials designed to resist chemicals, such as B-492 frozen surface polyester and B-481 chemical resistant polyester. The Brady IP® industrial label printer status monitor shows the printer and material information on the user's PC.
This ribbon is designed for use with Brady IP® Series Label Printer and BradyPrinter i5100 Industrial Label Printer.