Type J Temperature Controller, Ace Glass Incorporated
Supplier: Ace Glass
12126-24 12126-45
89076-516EA 2245.61 USD
89076-516 89076-518
Type J Temperature Controller, Ace Glass Incorporated
Controllers Temperature Controllers
Improved model temperature controller, for general lab use including mantles up to 22 L
Features two front mounted outlets, 16 segment ramp and soak function, fuzzy logic auto tune PID, type 'J' thermocouple, dual 4 digit LED display, universal TC jack for "mini" or "standard" plugs. Unit is downsized, compact and lightweight for bar mounting or benchtop operation. Temperature range of -120V, 50 to 800°C, input power 120VAC 50/60 Hz, 15 amps max., fused