Heat Gun Mite®, Ace Glass Incorporated
Supplier: Ace Glass
12072-20
89076-310EA 295.22 USD
89076-310
Heat Gun Mite®, Ace Glass Incorporated
Heaters Hot Air Dryers
Economical, compact, flameless heat gun designed for heat shrinkage as well as countless other industrial applications
Input is 120V, 5.4 amps. Supplied with 2.1m, three-conductor grounded cord. Unit consists of heat gun with silver nozzle and deflector adapter.