Varitemp® Heat Gun, Ace Glass Incorporated
Supplier: Ace Glass
12070-10
89076-308EA 592.19 USD
89076-308
Varitemp® Heat Gun, Ace Glass Incorporated
Heaters Hot Air Dryers
Heavy-duty, flameless heat gun features an exclusive locking electronic temperature control that enables the user to dial in the precise temperature for the job
The control locks in place to allow the same setting to be used repeatedly. Approximate temperature range at nozzle is ambient to 400°C (752°F). The heat gun features a 13/16" nozzle with a powerful turbo fan that is controlled by a three-way switch with guard. Reinforced ceramic heating element with double-jacketed heater housing for protection. Rugged, die-cast aluminum housing with externally replaceable carbon brushes. Supplied with a 243.8cm (8') neoprene cord with three-wire ground plug.