Heat Exchanger Coil Elbow Adapter, Ace Glass Incorporated
Supplier: Ace Glass
12067-02 12067-04 12067-06
89076-224EA 170.91 USD
89076-224 89076-226 89076-228
Heat Exchanger Coil Elbow Adapter, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
These are stainless steel elbow adapters for connecting a heat exchange coil tubing to a female pipe thread (FNPT).
  • Stainless steel
  • Makes a compression seal
  • Working pressure of 25 psig
Adapter fits over tubing and makes a compression seal. Working pressure of 25 psig. Complete item includes compression adapter, elbow, and pipe nipple.