Adjustable Chain Clamp, Ace Glass Incorporated
Supplier: Ace Glass
11079-38 11079-24 11079-40
89075-628EA 183.65 USD
89075-628 89083-212 89083-214
Adjustable Chain Clamp, Ace Glass Incorporated
Clamps General Purpose Clamps
The fast, sure way to secure large equipment to rods and lattices
Holds 76 to 165mm bottles, flasks, and large objects. Simply loop the steel chain around the object, attach to hook on movable slide, and tighten slide with large non-slip knob. Holds equipment firmly in place at any angle. You can easily vary the distance from the support rod.