Ultrasonic Reactor, Small Volume, Ace Glass Incorporated
Supplier: Ace Glass
9830-25
89075-524EA 18730.06 USD
89075-524
Ultrasonic Reactor, Small Volume, Ace Glass Incorporated
Laboratory Reactor Systems
Complete reaction assembly with parts necessary to perform mixing and reactions from 3 to 200 mL
Includes three borosilicate glass vessels, power supply with converter, 1/2" horn, 1/2" extender, 1/4" microtip, slide adapter and clamp.