Micro/Mini-Lab Cap Stopper, Ace Glass Incorporated
Supplier: Ace Glass
9730-10
89075-474EA 50.36 USD
89075-474
Micro/Mini-Lab Cap Stopper, Ace Glass Incorporated
Stoppers Glass Stoppers
With a standard taper 14/10 outer joint externally threaded for making O-Ring Cap Seal connection with mating inner joint
Supplied with cap and O-Ring.
Ordering information: For replacement caps, see 89075-248; for O-Rings, see 89060-994.