GC Collection Tube, Ace Glass Incorporated
Supplier: Ace Glass
9579-06
89075-192EA 59.46 USD
89075-192
GC Collection Tube, Ace Glass Incorporated
Tubes Test Tubes
Microscale gas chromatography collection tube for fraction collection followed by transfer of sample to Ace Glass' conical reaction 0.1 ml vial for storage.
A standard taper 5/5 inner joint on tube allows direct connection to heated exit port of GC column via Ace Glass' #150 tsainless steel connection adapter. Inner joint facilitates transfer since 0.1 ml vial has a standard taper 5/5 outer joint.