Micro/Mini-Lab Vacuum Take-Off Adapter with Side Joint, Ace Glass Incorporated
Supplier: Ace Glass
9565-05 9565-06
89075-150EA 108.63 USD
89075-150 89075-152
Micro/Mini-Lab Vacuum Take-Off Adapter with Side Joint, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
Used for Microscale vacuum distillations, especially for use with a spinning band microdistillation column.
  • Top joint is a combination cap, thread and standard taper joint
  • Bottom inner [ST] joint
  • [ST] outer side joint for vacuum connection or stopper
  • 105° angle
  • Cap and O-Ring included
The top joint is a combination cap, thread and standard taper joint to accommodate and seal other microscale apparatus. The O.D. of the top joint has an external thread to facilitate an O-Ring and open-top phenolic cap. The internal or I.D. at the top is a ground outer joint. Inner standard taper joint at other end and a standard taper 7/10 outer side joint for vacuum connection or stopper when using spinning band column technique. 105° angle. Supplied with cap and O-Ring.