Precision Ground Stirring Shaft, Ace Glass
Supplier: Ace Glass
9533-02
89075-076EA 189.35 USD
89075-076
Precision Ground Stirring Shaft, Ace Glass
Stirrers Stirring Shafts
Precision ground glass stirring shaft, 10 mm O.D., attached vanes
Vanes will fit through a standard taper 45/50 joint and are contoured for use with Ace series Mini-Lab flasks. Use with Trubore® bearings. Compatible with Ace mechanical seal and PTFE collet type bearings. Join to stirrer motor using series chucks.