Stirring Shaft, Borosilicate, Ace Glass Incorporated
Supplier: Ace Glass
9532-10
89075-074EA 309.35 USD
89075-074
Stirring Shaft, Borosilicate, Ace Glass Incorporated
Stirrers Stirring Shafts
Precision ground glass stirring shaft with attached vanes, 10 mm
For use when agitation and gas introduction are desired at the same time. Agitator will fit through a standard taper 45/50 joint. Designed for use with ACE Trubore® bearing and mini-lab flask heads. Introduced gas exits the hollow shaft within the agitator and produces a fine dispersion. Join to stirrer motor with series chucks.