Jacketed Flask with Polished Joint, Ace Glass Incorporated
Supplier: Ace Glass
9450-08
89074-576EA 501.4 USD
89074-576
Jacketed Flask with Polished Joint, Ace Glass Incorporated
Flasks Round Bottom Flasks
Jacketed version of standard flasks
Furnished with polished standard taper 45/50 outer joints so that it can be used with fitting heads.