Distillation Receiver, Use At Other Than Atmospheric Pressure, Ace Glass
Supplier: Ace Glass
9375-08
89074-456EA 759 USD
89074-456
Distillation Receiver, Use At Other Than Atmospheric Pressure, Ace Glass
Distillation Components Distillation Receivers
This receiver is designed for operation at other than atmospheric pressure
Vacuum is applied to the side arm with the T-bore stopcock. The secondary receiver is vented by means of the lower stopcock. The capacity from the bottom of the drip to the stopcock is 2.5 mL. The apparatus has a [ST] 14/20 outer joint at top, [ST] 14/20 inner joint at bottom. Hose connections are for use with 3/8-inch I.D. Both stopcocks are glass. Fabricated from borosilicate glass.