Distillation Receiver with Side Arm, Ace Glass
Supplier: Ace Glass
9373-06
89074-450EA 169.47 USD
89074-450
Distillation Receiver with Side Arm, Ace Glass
Distillation Components Distillation Receivers
The apparatus is graduated to 15mL in 1/10mL subdivisions, with the lower 0.5mL contained in a precision tip for better accuracy with extremely small samples
Fabricated from borosilicate glass.