Adapter, Vacuum Take-Off, Ace Glass Incorporated
Supplier: Ace Glass
5192-16 5192-33 5192-12 9124-05 9124-07
89051-370EA 135.04 USD
89051-370 89051-372 89051-374 89074-284 89074-288
Adapter, Vacuum Take-Off, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
The hose connection on the side of this adapter faces in the opposite direction of the top joint.
  • 105° angle outer joint
  • Side hose connection
  • Fabricated from borosilicate glass
Featuring an outer joint that is at an angle of 105°. The hose connection on the side faces in the opposite direction of the top joint. The stem on the 89074-284, 89074-288 is even with the joint bottom; on 89051-374, 89051-370 stem extends below the joint by 30mm.