WorkWipes® Reclaimed White Sweatshirt in Bag, PIG®
Supplier: New Pig
WIP553
89073-914EA 83.15 USD
89073-914
WorkWipes® Reclaimed White Sweatshirt in Bag, PIG®
Wipes Reusable Cleaning Cloths Cloths for General Applications
Eco- and cost-friendly rags are made from recycled, post-consumer cotton and cotton blend sweatshirt material.
  • Thick, absorbent rags are ideal for heavy-duty wiping
  • White color is ideal for use with solvents, which might cause colored rags to bleed
  • Never worry about contamination or poor performance
  • Hand-sorted and scanned to ensure use
  • Double scanned for metallic content
Strict material and size requirements mean you get more usable, high quality rags in every package. No useless rag pieces! All WorkWipes Reclaimed Rags are double scanned for metallic content and screened for printing when applicable, so you never have to worry about contamination or poor performance. Rags are visually hand-sorted and scanned to remove large printing to ensure they're usable.