DuraPrep™ Surgical Solution, 3M™
Supplier: Healthcare Products
MMM 8630
89073-162CS 362.48 USD
89073-162
DuraPrep™ Surgical Solution, 3M™
Skincare Soaps and Skin Cleansers
3M™ DuraPrep™ Surgical solution is a patient preoperative skin preparation, with applicator covers 450 inch.sq. (15×30"), ideal for large procedures such as Thoracic and CV, spine, total hip and knee, C-section, major abdominal.
  • Topical use
  • Cost effective
  • Broad-spectrum antimicrobial activity
Certifications: Meets AORN and CDC guidelines for the reduction of surgical site infections.