VWR® Conductive Bags, Black
Supplier: VWR International
89072-274
89072-274CS 205.6 USD
89072-274
VWR® Conductive Bags, Black
Bags Sample Bags
Constructed of heavy-duty 4 mm black carbon-impregnated low density polyethylene (LDPE) film to create an extra strong static barrier.
  • Polyethylene fabrication
  • Can be heat sealed, tied, stapled, or taped shut
Protects against electrostatic charge, chemicals, abrasion and humidity if heat-sealed.
Certifications: Meets specifications of MIL-P-82646, Rev. B.