VWR® Poly Sheeting / Pallet Top Sheets, 1.5 mil
Supplier: VWR International
89072-228
89072-228CS 493.62 USD
89072-228
VWR® Poly Sheeting / Pallet Top Sheets, 1.5 mil
Seals General Purpose Sealing Films
The 1.5 mm Low Density Polyethylene sheets easily tear off perforated rolls.
  • Low-density polyethylene construction
  • Clear for quick identification
  • Meets FDA/USDA specifications
Made of 100% virgin LDPE resin that meets FDA/USDA specifications. Use as pallet top covers, drop cloths, or dust covers. Wound on a 3" core with 1" diameter core plug. Folds may occur on either side of the roll.
Certifications: Meet FDA/USDA specifications.