Radiation Alert® PEN Dosimeters, S.E. International
Supplier: SE International
PEN-DOSIMETER
89068-906EA 695.59 USD
89068-906
Radiation Alert® PEN Dosimeters, S.E. International
Radiation Meters
The advanced PEN direct reading dosimeters are rugged, precision instruments
  • Measures and directly reads, at any time, accumulated dose of gamma and x-ray exposure
  • Hermetically sealed and immersion proof
  • Available in several scale ranges, 200mR, 500mR, 2R, 5R, 20R, 2mSv, 5mSv (sold separately)
A sturdy metal clip attaches the dosimeter to a pocket or any object to monitor total radiation exposure.
Ordering information: Contact your Avantor representative for available scale ranges. Consult the datasheet for more information.