Esteem® Stretchy Nitrile Ambi Gloves, Cardinal
Supplier: Avantor
89068-892CS 1216.57 USD
89068-892 89068-894 89068-888
Esteem® Stretchy Nitrile Ambi Gloves, Cardinal
Gloves
Ambidextrous synthetic nitrile gloves have beaded cuffs and textured fingers.
  • Gloves are powder-and latex-free
Unique stretchy nitrile formulation provides the protection of nitrile while also offering comfort and flexibility.
Ordering information: Gloves are flat packed into double poly bags, heat-sealed, and placed into a poly case liner.