Super Challenger™ Enrichment Device, Bio-Serv
Supplier: Bio-Serv
K3100-S
89067-782EA 72.26 USD
89067-782
Super Challenger™ Enrichment Device, Bio-Serv
Animal Supplies Animal Environmental Enrichment
Made of sturdy polyethylene with a 10" stainless steel adjustable chain
Internal disk provides additional challenge. Hole size accommodates larger treats. Hang from outside of cage for easy filling. Cagewasher safe.