Shake-A-Treat Enrichment Device, Bio-Serv
Supplier: Bio-Serv
K3350
89067-780EA 84.9 USD
89067-780
Shake-A-Treat Enrichment Device, Bio-Serv
Animal Supplies Animal Environmental Enrichment
Made of sturdy, clear polycarbonate and stainless steel
Interactive foraging device to promote mental stimulation. Easy top loading. No need to disassemble. Vary the level of difficulty by using different sized treats up to 1/2" diameter. Easy-to-clean 2-piece construction. Hang from cage by an adjustable welded link stainless steel chain with quick link (overall chain length is 10"). Autoclavable. Cagewasher safe.