Chain Assembly for Enrichment Devices, Round Bead, Bio-Serv
Supplier: Bio-Serv
K9004
89067-736EA 3.32 USD
89067-736
Chain Assembly for Enrichment Devices, Round Bead, Bio-Serv
Animal Supplies Animal Environmental Enrichment
Made entirely of stainless steel
1/8" round beads. A convenient way to suspend enrichment products. Autoclavable. Cagewasher safe. Assembly required.