Fenbendazole Rodent Diet, Grain-Based, Bio-Serv

Supplier: Bio-Serv
S3867
89067-465BX 309.52 USD
89067-465
Fenbendazole Rodent Diet, Grain-Based, Bio-Serv
Animal Supplies Animal Feed
A proven effective and safe treatment for pinworms.

  • Nutritionally complete grain-based rodent diet containing 150 mg of Fenbendazole per kg per diet
  • Sterilized by gamma irradiation
  • Nutritionally assayed
  • Shelf Life: 6 months, cool dry storage

Ingredients: Ground Corn, Dehulled Soybean Meal, Dried Beet Pulp, Fish Meal, Ground Oats, Brewers Dried Yeast, Cane Molasses, Dehydrated Alfalfa Meal, Dried Whey, Wheat Germ, Porcine Animal Fat (preserved with BHA), Porcine Meat Meal, Wheat Middlings, Mineral Mix, Vitamin Mix, DL-Methionine, Choline Chloride, Fenbendazole.

Caloric value: 3.35 kcal/g
Nutritional ash content: 7.0%
Nutritional carbohydrate content: 48.7%
Nutritional fat content: 5.0%
Nutritional fibre content: 5.1%
Nutritional moisture content: <10%
Nutritional protein content: 23.9%
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR