SCIDS MD's™ Severe Combined Immunodeficiency Mouse Tablets, Sterile, Bio-Serv

Supplier: Bio-Serv
S0443
89067-458PK 107.97 USD
89067-458
SCIDS MD's™ Severe Combined Immunodeficiency Mouse Tablets, Sterile, Bio-Serv
Animal Supplies Animal Feed
An effective method to reduce the hazards of respiratory problems in immunocompromised mice

  • Bacon-flavored tablets are highly palatable, nutritionally complete, and grain-based
  • More convenient and minimizes risk of dehydration compared to using medicated water
  • Direct compression process eliminates drug degradation
  • Sterilized by gamma irradiation
  • Nutritionally assayed
  • Shelf Life: 12 months, cool dry storage.

Ingredients: Ground Corn, Banana Flakes, Dehulled Soybean Meal, Corn Gluten Meal, Ground Wheat, Corn Gluten Feed, Fish Meal, Dehydrated Alfalfa Meal, Casein, Dried Whey, Sucrose, Fructose, Dextrose, Dried Beet Pulp, Soybean Oil, Porcine Animal Fat (Preserved with BHA), Dried Brewers Yeast, Mineral Mix, Vitamin Mix, Magnesium Stearate, Calcium Silicate, Bacon Flavor, DL-Methionine, Choline Chloride, Sodium Propionate, Ascorbic Acid, Kaolin, Sulfamethoxazole, Trimethoprim.

Each bacon-flavored, scored tablet contains 60 mg Sulfamethoxazole and 12 mg Trimethoprim. Mouse average dose: One 5 g tab/20 g of body weight. To administer, place on cage floor for easy access.

Caloric value: 3.39 kcal/g
Nutritional ash content: 7.8%
Nutritional carbohydrate content: 54.0%
Nutritional fat content: 4.3%
Nutritional fibre content: 4.0%
Nutritional moisture content: <10%
Nutritional protein content: 21.3%
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR