Mouse Helicobacter MD's™ 4 Drug Combo Tablet, Sterile, Bio-Serv

Supplier: Bio-Serv
S05723
89067-452PK 101.69 USD
89067-452
Mouse Helicobacter MD's™ 4 Drug Combo Tablet, Sterile, Bio-Serv
Animal Supplies Animal Feed
The most effective oral treatment against a broad spectrum of Helicobacter strains

  • Highly palatable, bacon-flavored, nutritionally complete diet. Used to treat breeders or quarantined mice
  • Alternative method of treatment to cesarean rederivation
  • Comprehesive husbandry plan must be employed for success; contact Bio-Serv for details
  • Nutritionally assayed
  • Shelf Life: 12 months, cool dry storage

Ingredients: Ground Corn, Dehulled Soybean Meal, Dried Beet Pulp, Fish Meal, Ground Oats, Brewers Dried Yeast, Cane Molasses, Dehydrated Alfalfa Meal, Dried Whey, Wheat Germ, Porcine Animal Fat (preserved with BHA), Porcine Meat Meal, Wheat Middlings, Mineral Mix, Vitamin Mix, Bacon Flavor, DL-Methionine, Choline Chloride, Omeprazole, Metronidazole, Amoxicillin, Clarithromycin.

Each 5 g Mouse Helicobacter MD contains 3.00 mg of Amoxicillin, 0.50 mg of Clarithromycin, 1.00 mg of Metronidazole and 0.02 mg of Omeprazole. Mouse average dose: One 5 g tablet/50 g of body weight.

Caloric value: 3.39 kcal/g
Nutritional ash content: 7.8%
Nutritional carbohydrate content: 54.0%
Nutritional fat content: 4.3%
Nutritional fibre content: 4.0%
Nutritional moisture content: <10%
Nutritional protein content: 21.3%
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR